Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gprc5c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109713.1 | NP_001103183.1 | 4 - 5 | 1585 | 146 | |
GenBank mRNA | BC151953.1 | - | 4 - 5 | 1585 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |