Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ckma | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130932.2 | NP_571007.2 | 3 - 4 | 456 | 136 | |
GenBank mRNA | AF134851.1 | - | 3 - 4 | 382 | 136 | |
AY036972.1 | - | 3 - 4 | 410 | 136 | ||
BC056706.1 | - | 3 - 4 | 428 | 136 | ||
BC065844.1 | - | 3 - 4 | 428 | 136 | ||
EH439264.1 | - | 3 - 4 | 448 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Danio rerio Inventoried Probe spans exons |