Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gdf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130948.1 | NP_571023.1 | 1 - 2 | 267 | 87 | |
XM_017351595.2 | XP_017207084.1 | 4 - 5 | 741 | 87 | ||
XM_021467105.1 | XP_021322780.1 | 2 - 3 | 772 | 87 | ||
GenBank mRNA | BC085547.1 | - | 1 - 2 | 267 | 87 | |
BC164172.1 | - | 1 - 2 | 290 | 87 | ||
U00931.1 | - | 1 - 2 | 267 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Inventoried Probe spans exons |