Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: wnt8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130959.2 | NP_571034.1 | 4 - 5 | 396 | 92 | |
GenBank mRNA | KM655728.1 | - | 4 - 5 | 367 | 92 | |
U10870.1 | - | 4 - 5 | 400 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Danio rerio Probe spans exons |