Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rplp0 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131580.2 | NP_571655.2 | 7 - 8 | 865 | 132 | |
GenBank mRNA | AF134852.1 | - | 7 - 8 | 874 | 132 | |
BC049058.1 | - | 7 - 8 | 877 | 132 | ||
BC062854.1 | - | 7 - 8 | 865 | 132 | ||
BC146738.1 | - | 7 - 8 | 917 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Inventoried Probe spans exons |