Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dhfr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131775.1 | NP_571850.1 | 3 - 4 | 330 | 94 | |
GenBank mRNA | AY026507.1 | - | 3 - 4 | 250 | 94 | |
BC071330.1 | - | 3 - 4 | 330 | 94 | ||
EU145591.1 | - | 3 - 4 | 247 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |