Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: efnb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131805.1 | NP_571880.1 | 3 - 4 | 490 | 118 | |
GenBank mRNA | AF375224.1 | - | 3 - 4 | 490 | 118 | |
BC163323.1 | - | 3 - 4 | 586 | 118 | ||
BC163326.1 | - | 3 - 4 | 586 | 118 | ||
CU638799.1 | - | 3 - 4 | 1386 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |