Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tnnt2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_152893.1 | NP_690853.1 | 11 - 12 | 630 | 103 | |
XM_005161930.4 | XP_005161987.1 | 10 - 11 | 669 | 103 | ||
XM_021469840.1 | XP_021325515.1 | 10 - 11 | 694 | 103 | ||
GenBank mRNA | AF282384.1 | - | 11 - 12 | 630 | 103 | |
AF434187.1 | - | 11 - 12 | 630 | 103 | ||
BC108081.1 | - | 11 - 12 | 633 | 103 | ||
BC153503.1 | - | 11 - 12 | 757 | 103 | ||
BC153911.1 | - | 11 - 12 | 622 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Inventoried Probe spans exons |