Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: spns1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153663.2 | NP_705949.2 | 11 - 12 | 1508 | 87 | |
XM_005163832.4 | XP_005163889.1 | 11 - 12 | 1522 | 87 | ||
GenBank mRNA | AF465772.1 | - | 11 - 12 | 87 | ||
BC048024.1 | - | 11 - 12 | 87 | |||
BC165625.1 | - | 10 - 11 | 87 | |||
CF999294.1 | - | 3 - 4 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |