Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: spns1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153663.2 | NP_705949.2 | 6 - 7 | 859 | 92 | |
XM_005163832.4 | XP_005163889.1 | 6 - 7 | 873 | 92 | ||
GenBank mRNA | AF465772.1 | - | 6 - 7 | 815 | 92 | |
BC048024.1 | - | 6 - 7 | 814 | 92 | ||
BC165625.1 | - | 5 - 6 | 659 | 92 | ||
CK027241.1 | - | 1 - 2 | 69 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |