Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tbx15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001328433.1 | NP_001315362.1 | 4 - 5 | 999 | 128 | |
XM_021478599.1 | XP_021334274.1 | 4 - 5 | 851 | 128 | ||
GenBank mRNA | AF448504.1 | - | 4 - 5 | 965 | 128 | |
BC128812.1 | - | 4 - 5 | 961 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |