Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: u2af1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177479.2 | NP_803432.1 | 3 - 4 | 200 | 95 | |
GenBank mRNA | AF506202.1 | - | 3 - 4 | 200 | 95 | |
BC098601.1 | - | 3 - 4 | 227 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |