Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: drd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183067.1 | NP_898890.1 | 1 - 2 | 320 | 70 | |
XM_005162673.4 | XP_005162730.1 | 2 - 3 | 1080 | 70 | ||
XM_021470111.1 | XP_021325786.1 | 2 - 3 | 421 | 70 | ||
GenBank mRNA | AY183455.1 | - | 1 - 2 | 320 | 70 | |
BC076352.1 | - | 2 - 3 | 593 | 70 | ||
BC165727.1 | - | 1 - 2 | 313 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |