Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mibp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198877.1 | NP_942578.1 | 3 - 4 | 371 | 124 | |
GenBank mRNA | BC047858.1 | - | 3 - 4 | 371 | 124 | |
BC066586.1 | - | 3 - 4 | 319 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |