Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: waca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199660.1 | NP_955954.1 | 3 - 4 | 395 | 81 | |
XM_005156197.2 | XP_005156254.1 | 3 - 4 | 491 | 81 | ||
XM_009306588.2 | XP_009304863.1 | 3 - 4 | 491 | 81 | ||
GenBank mRNA | AY576994.1 | - | 3 - 4 | 400 | 81 | |
BC048888.1 | - | 3 - 4 | 395 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |