Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gosr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199688.1 | NP_955982.1 | 5 - 6 | 514 | 100 | |
GenBank mRNA | BC053235.1 | - | 5 - 6 | 514 | 100 | |
BC165142.1 | - | 5 - 6 | 512 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |