Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: upp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200144.1 | NP_956438.1 | 1 - 2 | 242 | 85 | |
GenBank mRNA | BC044525.1 | - | 1 - 2 | 242 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |