Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dnajb6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200305.1 | NP_956599.1 | 2 - 3 | 374 | 92 | |
XM_009303365.3 | XP_009301640.1 | 2 - 3 | 444 | 92 | ||
XM_009303366.2 | XP_009301641.1 | 2 - 3 | 459 | 92 | ||
XM_021477435.1 | XP_021333110.1 | 2 - 3 | 459 | 92 | ||
GenBank mRNA | BC049536.1 | - | 2 - 3 | 374 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |