Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc2a12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200538.1 | NP_956832.1 | 4 - 5 | 1658 | 93 | |
XM_005162304.4 | XP_005162361.1 | 5 - 6 | 1852 | 93 | ||
GenBank mRNA | BC056306.1 | - | 4 - 5 | 1658 | 93 | |
BC067616.1 | - | 4 - 5 | 1666 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |