Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc2a12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200538.1 | NP_956832.1 | 5 - 6 | 1790 | 137 | |
XM_005162304.4 | XP_005162361.1 | 6 - 7 | 1984 | 137 | ||
GenBank mRNA | BC056306.1 | - | 5 - 6 | 1790 | 137 | |
BC067616.1 | - | 5 - 6 | 1798 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |