Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: arl2bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201001.2 | NP_957295.2 | 5 - 6 | 405 | 90 | |
GenBank mRNA | AB128034.1 | - | 4 - 5 | 295 | 90 | |
BC054678.1 | - | 5 - 6 | 418 | 90 | ||
BC066483.1 | - | 5 - 6 | 357 | 90 | ||
EH541682.1 | - | 5 - 6 | 428 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |