Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rapgef1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021468784.1 | XP_021324459.1 | 11 - 12 | 2157 | 84 | |
GenBank mRNA | BC049305.1 | - | 10 - 11 | 1955 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |