Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: flot1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201456.1 | NP_958864.1 | 4 - 5 | 323 | 70 | |
GenBank mRNA | AY398349.1 | - | 4 - 5 | 323 | 70 | |
BC049425.1 | - | 4 - 5 | 291 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |