Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cga | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205687.2 | NP_991250.1 | 1 - 2 | 96 | 118 | |
XM_005169876.4 | XP_005169933.1 | 2 - 3 | 329 | 118 | ||
GenBank mRNA | AY424306.1 | - | 2 - 3 | 135 | 118 | |
AY522553.1 | - | 2 - 3 | 118 | |||
BC116611.1 | - | 1 - 2 | 96 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |