Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: atp6v0a1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212672.1 | NP_997837.1 | 8 - 9 | 795 | 97 | |
XM_005163798.3 | XP_005163855.1 | 8 - 9 | 907 | 97 | ||
GenBank mRNA | BC066692.1 | - | 8 - 9 | 795 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |