Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mttp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212970.2 | NP_998135.2 | 5 - 6 | 787 | 95 | |
GenBank mRNA | AJ428850.1 | - | 5 - 6 | 680 | 95 | |
BC135020.1 | - | 5 - 6 | 764 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |