Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eif4bb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214718.1 | NP_999883.1 | 6 - 7 | 702 | 68 | |
XM_005166014.4 | XP_005166071.1 | 6 - 7 | 724 | 68 | ||
XM_021476791.1 | XP_021332466.1 | 6 - 7 | 723 | 68 | ||
GenBank mRNA | AY576997.1 | - | 6 - 7 | 702 | 68 | |
BC066675.1 | - | 5 - 6 | 679 | 68 | ||
BC086699.1 | - | 6 - 7 | 586 | 68 | ||
BC164129.1 | - | 5 - 6 | 613 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |