Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: arhgef18a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214818.1 | NP_999983.1 | 11 - 12 | 1544 | 77 | |
XM_005171284.4 | XP_005171341.1 | 10 - 11 | 1587 | 77 | ||
XM_009298637.3 | XP_009296912.1 | 12 - 13 | 1651 | 77 | ||
XM_021479667.1 | XP_021335342.1 | 11 - 12 | 1624 | 77 | ||
GenBank mRNA | BC070002.1 | - | 11 - 12 | 1544 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |