Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eif2b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002060.1 | NP_001002060.1 | 5 - 6 | 669 | 65 | |
GenBank mRNA | BC071346.1 | - | 5 - 6 | 669 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Endogenous Controls Danio rerio Probe spans exons |