Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: trim55a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002358.1 | NP_001002358.1 | 6 - 7 | 977 | 99 | |
GenBank mRNA | BC075897.1 | - | 6 - 7 | 977 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |