Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vps26bl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002415.2 | NP_001002415.1 | 3 - 4 | 772 | 137 | |
GenBank mRNA | BC076160.1 | - | 3 - 4 | 774 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |