Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc25a36a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002667.1 | NP_001002667.1 | 2 - 3 | 562 | 76 | |
GenBank mRNA | BC076521.1 | - | 2 - 3 | 562 | 76 | |
BC164094.1 | - | 2 - 3 | 234 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |