Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rfc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003862.1 | NP_001003862.1 | 8 - 9 | 803 | 85 | |
GenBank mRNA | AY648755.1 | - | 8 - 9 | 803 | 85 | |
BC095222.1 | - | 8 - 9 | 806 | 85 | ||
BC165456.1 | - | 8 - 9 | 806 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |