Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: myo6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004110.1 | NP_001004110.1 | 8 - 9 | 822 | 62 | |
XM_005158836.4 | XP_005158893.1 | 9 - 10 | 1076 | 62 | ||
XM_009293188.3 | XP_009291463.1 | 9 - 10 | 1076 | 62 | ||
XM_021467147.1 | XP_021322822.1 | 9 - 10 | 1080 | 62 | ||
XM_021467148.1 | XP_021322823.1 | 9 - 10 | 1079 | 62 | ||
GenBank mRNA | AY618701.1 | - | 8 - 9 | 822 | 62 | |
AY691328.1 | - | 9 - 10 | 975 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Danio rerio Probe spans exons |