Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: phf20b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004114.1 | NP_001004114.1 | 5 - 6 | 519 | 86 | |
XM_005161993.4 | XP_005162050.1 | 5 - 6 | 582 | 86 | ||
GenBank mRNA | BC080250.1 | - | 5 - 6 | 519 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |