Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rassf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004550.1 | NP_001004550.1 | 1 - 2 | 333 | 114 | |
XM_005173951.4 | XP_005174008.1 | 2 - 3 | 975 | 114 | ||
XM_017353113.2 | XP_017208602.1 | 2 - 3 | 694 | 114 | ||
GenBank mRNA | BC081661.1 | - | 1 - 2 | 333 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |