Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: azin1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007159.2 | NP_001007160.2 | 6 - 7 | 944 | 94 | |
GenBank mRNA | AB069858.1 | - | 4 - 5 | 578 | 94 | |
BC061960.1 | - | 4 - 5 | 600 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |