Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: wnt3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007185.1 | NP_001007186.1 | 2 - 3 | 311 | 76 | |
GenBank mRNA | AY613787.1 | - | 2 - 3 | 311 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Danio rerio Probe spans exons |