Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cd2ap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008583.2 | NP_001008583.2 | 11 - 12 | 1383 | 56 | |
XM_005160684.4 | XP_005160741.1 | 10 - 11 | 1014 | 56 | ||
XM_009294709.3 | XP_009292984.1 | 11 - 12 | 1414 | 56 | ||
GenBank mRNA | DT081164.1 | - | 6 - 7 | 503 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Danio rerio Probe spans exons |