Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cd2ap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008583.2 | NP_001008583.2 | 16 - 17 | 1941 | 82 | |
XM_005160684.4 | XP_005160741.1 | 15 - 16 | 1572 | 82 | ||
XM_009294709.3 | XP_009292984.1 | 16 - 17 | 1966 | 82 | ||
GenBank mRNA | DT075242.1 | - | 5 - 6 | 538 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Danio rerio Probe spans exons |