Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rps24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012316.1 | NP_001012316.1 | 3 - 4 | 313 | 77 | |
XM_005156806.4 | XP_005156863.1 | 3 - 4 | 391 | 77 | ||
XM_005156807.4 | XP_005156864.1 | 3 - 4 | 391 | 77 | ||
XM_021480547.1 | XP_021336222.1 | 3 - 4 | 519 | 77 | ||
GenBank mRNA | BC081494.1 | - | 3 - 4 | 313 | 77 | |
BC094973.1 | - | 3 - 4 | 313 | 77 | ||
BC165902.1 | - | 2 - 3 | 314 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |