Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: b3gat1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015066.2 | NP_001015066.1 | 4 - 5 | 1056 | 63 | |
XM_005173645.4 | XP_005173702.1 | 8 - 9 | 1984 | 63 | ||
XM_005173647.4 | XP_005173704.1 | 7 - 8 | 1910 | 63 | ||
GenBank mRNA | BC133855.1 | - | 4 - 5 | 1056 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |