Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hsh2d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017541.2 | NP_001017541.2 | 2 - 3 | 418 | 91 | |
GenBank mRNA | BC092186.1 | - | 2 - 3 | 422 | 91 | |
BC164408.1 | - | 1 - 2 | 158 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |