Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hsh2d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017541.2 | NP_001017541.2 | 5 - 6 | 900 | 63 | |
GenBank mRNA | BC092186.1 | - | 5 - 6 | 904 | 63 | |
BC164408.1 | - | 4 - 5 | 640 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |