Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: asb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017628.1 | NP_001017628.1 | 1 - 2 | 336 | 68 | |
XM_005159719.4 | XP_005159776.1 | 1 - 2 | 938 | 68 | ||
GenBank mRNA | BC092822.1 | - | 1 - 2 | 336 | 68 | |
BC165810.1 | - | 1 - 2 | 255 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Inventoried Probe spans exons |