Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: akr1a1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017640.1 | NP_001017640.1 | 5 - 6 | 621 | 112 | |
XM_005166285.3 | XP_005166342.1 | 5 - 6 | 686 | 112 | ||
XM_005166286.4 | XP_005166343.1 | 5 - 6 | 644 | 112 | ||
XM_005166287.3 | XP_005166344.1 | 5 - 6 | 755 | 112 | ||
XM_005166289.4 | XP_005166346.1 | 5 - 6 | 735 | 112 | ||
GenBank mRNA | BC092808.1 | - | 5 - 6 | 621 | 112 | |
BC155150.1 | - | 5 - 6 | 600 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |