Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lrrc8aa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029949.1 | NP_001025120.1 | 2 - 3 | 796 | 59 | |
GenBank mRNA | BC097172.1 | - | 2 - 3 | 796 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |