Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tpra1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030251.1 | NP_001025422.1 | 7 - 8 | 806 | 153 | |
GenBank mRNA | BC097163.1 | - | 7 - 8 | 806 | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |