Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: alg13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031838.1 | NP_001027008.1 | 14 - 15 | 1373 | 58 | |
XM_005161414.4 | XP_005161471.1 | 13 - 14 | 1468 | 58 | ||
XM_005161417.4 | XP_005161474.1 | 14 - 15 | 1366 | 58 | ||
GenBank mRNA | AY918066.1 | - | 14 - 15 | 1373 | 58 | |
BC057511.1 | - | 13 - 14 | 1354 | 58 | ||
BC133108.1 | - | 14 - 15 | 1280 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |