Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pmelb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033918.2 | NP_001029090.2 | 7 - 8 | 1201 | 120 | |
XM_005162378.4 | XP_005162435.1 | 7 - 8 | 1243 | 120 | ||
GenBank mRNA | AY864065.1 | - | 7 - 8 | 1171 | 120 | |
BC116462.1 | - | 7 - 8 | 1201 | 120 | ||
KM655726.1 | - | 7 - 8 | 1171 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |